Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ion Total RNA-Seq Kit v2

Catalog No. 4479789
Encompass_Preferred
Change view
Click to view available options
No. of Reactions:
48 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
44-797-89 48 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-797-89 Supplier LIFE TECHNOLOGIES Supplier No. 4479789
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Content And Storage Store at -20°C:
1 tube, 10X RT Buffer 48 reactions
1 tube, 2.5 mM dNTP Mix 48 reactions
1 tube, Platinum HiFi MasterMix 48 reactions
1 tube, 10x Superscript Enzyme Mix 48 reactions
1 tube, Ion 3'-PCR-primer v2 48 reactions
1 tube, Ion 5'-PCR-primer v2 48 reactions
1 tube, Ion 5'-PCR-primer v2 48 reactions
1 tube, Ion RT Primer v2 48 reactions
1 tube, Ion Adaptor Mix v2 48 reactions

Store at 4°C:
1 bottle, Binding Soluntion Concentrated 48 reactions
1 bottle, Wash Soln Concentrated 48 reactions
1 bottle, Nucleic Acid Binding Beads 48 reactions
Sample Type RNA
For Use With (Equipment) Ion GeneStudio™ S5, Ion PGM™, Ion Proton™ Systems
No. of Reactions 48 Reactions
Libraries cDNA Library
Product Line Ion Torrent
Quantity 48 reactions
Sequencing Type Transcriptome Sequencing
Starting Material Amount For RNA (rRNA-depleted): 1-100 ng mRNA
For whole transcriptome: ≥100 ng total RNA
Workflow Step NGS Library Generation
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.