Learn More
Thermo Scientific Ion Transfer Tube
Supplier:  Thermo Scientific 9713320001
Encompass
Ion transfer tube™ efficiently transfers ionized molecules between mass spectrometry ion source & mass analyzer under vacuum for ion transport, ion detection & analysis of ionized analytes.
Catalog No. NC9901670