Learn More
Electron Microscopy Sciences Rubis Ion Tweezers Style OO, 115 mm OAL
SDP
Catalog No. 5024070
Encompass_Preferred
This innovation has  resulted in a special coating surface on the tweezers which releases silver ions. These ions act as an anti-bacterial agent which helps protect the user from contamination of any pathogens. Unlike other methods, the use of ion technology does not give rise to any resistance. Consequently, it represents a risk-free method of guaranteeing hygiene, even at workstations where instruments are used by several different people.
Made from finest stainless steel grade, these tweezers are perfect companions for your daily use in the laboratory.
We recommend that safety gloves still be used whenever handling any infectious materials.
OAL: 115 mm
Catalog No. 50-240-70 Supplier Electron Microscopy Sciences Supplier No. 78146OO
This innovation has  resulted in a special coating surface on the tweezers which releases silver ions. These ions act as an anti-bacterial agent which helps protect the user from contamination of any pathogens. Unlike other methods, the use of ion technology does not give rise to any resistance. Consequently, it represents a risk-free method of guaranteeing hygiene, even at workstations where instruments are used by several different people.
Made from finest stainless steel grade, these tweezers are perfect companions for your daily use in the laboratory.
We recommend that safety gloves still be used whenever handling any infectious materials.
OAL: 115 mm