Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ion Torrent™ Ion Xpress™ RNA-Seq Barcode 1-16 Kit

Catalog No. 4475485
Encompass_Preferred
Change view
Click to view available options
Quantity:
160 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-754-85 160 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-754-85 Supplier Ion Torrent™ Supplier No. 4475485
Add to Cart
Add to Cart

The new Ion Xpress™ RNA-Seq Barcode 1-16 Kit provides a set of 16 unique barcode primers for sequencing RNA on the Ion Torrent PGM™ semiconductor sequencer.

The new Ion Xpress™ RNA-Seq Barcode 1-16 Kit provides a set of 16 unique barcode primers for sequencing RNA on the Ion Torrent PGM™ semiconductor sequencer. When used in combination with the Ion Total RNA-Seq Kit v2 (Cat. No. 4475936), the Ion Xpress™ RNA-Seq Barcode 1-16 Kit enables users to pool up to 16 fragment libraries prior to template preparation and then conduct multiplexed sequencing analysis, simplifying the Ion semiconductor sequencing workflow for small and whole transcriptome RNA.

Features and Benefits:

  • Efficient multiplexing-enables multiplexing of up to 16 fragment libraries on a single sequencing chip
  • Both sequence- and flow-optimized for equal representation of all barcodes in a pool
  • Multiplexing decreases the cost and the handling requirements of template preparation and sequencing
  • Robust error correction built in by design for added confidence in sample identification

Efficient Multiplexing with Barcode Primers Enables Higher Throughput Sample Sequencing

Multiplexing with barcoded libraries leads to more cost-effective runs by permitting up to 16 samples per run, significantly decreasing the cost and handling requirements of template preparation and sequencing.

Sequence- and Flow-Optimized for Increased Performance and Efficiency

Barcode primers contain sequences that are optimized to provide equal representation of all barcodes in a pool, require a minimum number of flows to interrogate, and necessitate only 20 bases of barcode and adaptor sequencing, which results in more economical multiplexed sequencing runs.

Robust Error Correction Built In By Design

Built-in error correction significantly minimizes the possibility of incorrect sample assignment.

The Ion Xpress™ RNA-Seq Barcode 01-16 Kit is designed to create 10 multiplexed libraries per barcode or a total of 160 multiplexed libraries.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • Ion Xpress™ RNA BC 01–BC 16, 16 x 12 μL tubes, store at 20°C
• Ion Xpress™ RNA 3' Barcode Primer, 1 x 192 μL, store at 20°C
For Use With (Equipment) Ion GeneStudio™ S5 Series Sequencer, Ion Chef™ System
No. of Reactions 160
Quantity 160 Reactions
Shipping Condition Dry Ice
Workflow Step Library Barcoding

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.