Learn More
IONBENCH CORPORATION IonBench for LCMS with dry pump Dimensions: W51" x D38" x H34" (W130 x D97 x H86 cm )
Catalog No. 5000150573
Encompass_Preferred
IonBench for LCMS with dry pump. Dimensions: W51" x D38" x H34" (W130 x D97 x H86 cm ) Bundle Includes: TRESPA_WS_MS Countertop upgraded to Trespa Toplab, chemically resistant. DR3 Bank of three drawers. PC1 CPU holder to be fixed on one side of the bench Installable right or left. ARM1 Pack of a single monitor arm and sliding keyboard. ELECUS Power strip, North American 8 outlets fixed behind the bench, NEMA5-15 outlets. LAT-SOLV Side fixed container for solvent waste bottles Dim : W25xD38xH10cm (W9.8"xD14.9"xD3.9")
Catalog No. 50-001-50573 Supplier IONBENCH CORPORATION Supplier No. BCH5138INCHDRY
Add to Cart
Add to Cart
Request a Quote
IonBench for LCMS with dry pump. Dimensions: W51" x D38" x H34" (W130 x D97 x H86 cm ) Bundle Includes: TRESPA_WS_MS Countertop upgraded to Trespa Toplab, chemically resistant. DR3 Bank of three drawers. PC1 CPU holder to be fixed on one side of the bench Installable right or left. ARM1 Pack of a single monitor arm and sliding keyboard. ELECUS Power strip, North American 8 outlets fixed behind the bench, NEMA5-15 outlets. LAT-SOLV Side fixed container for solvent waste bottles Dim : W25xD38xH10cm (W9.8"xD14.9"xD3.9")