Learn More
IONBENCH CORPORATION IonBench for LCMS with dry pump Dimensions:W75" x D38" x H34" ( W190 x D97 x H86 cm)
Catalog No. 5000150569
Encompass_Preferred
IonBench for LCMS with dry pump Dimensions:W75" x D38" x H34" ( W190 x D97 x H86cm) Included: TRESPA_WS_MS Countertop upgraded to Trespa Toplab, chemically resistant, DR3 Bank of three drawers, ARM1 Pack of a single monitor arm and sliding keyboard, PC1 CPU holder to be fixed on one side of the bench Installable right or left, ELECUS Power strip, North American 8 outlets fixed behind the bench, NEMA5-15 outlets, SOLV Container for solvent waste bottles
Catalog No. 50-001-50569 Supplier IONBENCH CORPORATION Supplier No. BCH7538INCHDRY
Add to Cart
Add to Cart
Request a Quote
IonBench for LCMS with dry pump Dimensions:W75" x D38" x H34" ( W190 x D97 x H86cm) Included: TRESPA_WS_MS Countertop upgraded to Trespa Toplab, chemically resistant, DR3 Bank of three drawers, ARM1 Pack of a single monitor arm and sliding keyboard, PC1 CPU holder to be fixed on one side of the bench Installable right or left, ELECUS Power strip, North American 8 outlets fixed behind the bench, NEMA5-15 outlets, SOLV Container for solvent waste bottles