Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters IonHance CX-MS pH Concentrate A, 10X Concentrate, MS Certified, 100 mL

Catalog No. 502038967 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This 10X liquid concentrate is designed for creating gradients in LC/MS analysis of monoclonal antibody (mAb) charge variant separations. It offers high purity to limit background contribution and features modifiers for reversed-phase separations of proteins, peptides, and small compounds, improving sensitivity, retentivity, and peak shape.

  • Optimized for LC/MS analysis of mAb charge variant separations.
  • Designed for high-purity mobile-phase additives.
  • Employed for reversed-phase separations of proteins, peptides, and small compounds.
  • Achieves minimal metal content and good quality standards.
  • Improves sensitivity, retentivity, and peak shape.
  • Separation mode: Ion Exchange Chromatography.
  • Detection method: LCMS.
  • Formulation: Liquid.
  • Volume: 100 mL.
  • Shelf life: 12 months.
  • Shelf life upon reconstitution: 1 week.

Catalog No. 50-203-8967 Supplier Waters Corp Supplier No. 186009280
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This 10X liquid concentrate is designed for creating gradients in LC/MS analysis of monoclonal antibody (mAb) charge variant separations. It offers high purity to limit background contribution and features modifiers for reversed-phase separations of proteins, peptides, and small compounds, improving sensitivity, retentivity, and peak shape.

  • Optimized for LC/MS analysis of mAb charge variant separations.
  • Designed for high-purity mobile-phase additives.
  • Employed for reversed-phase separations of proteins, peptides, and small compounds.
  • Achieves minimal metal content and good quality standards.
  • Improves sensitivity, retentivity, and peak shape.
  • Separation mode: Ion Exchange Chromatography.
  • Detection method: LCMS.
  • Formulation: Liquid.
  • Volume: 100 mL.
  • Shelf life: 12 months.
  • Shelf life upon reconstitution: 1 week.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.