Learn More
Waters Corp Waters Corporation Concentrate, IonHance CX-MS pH, pH 8.5, 10X Concentrate
Catalog No. 502038968 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This 10X concentrate was developed in conjunction with BioResolve SCX columns and is optimized for creating a gradient for LC/MS analysis of mAb charge variant separations. It is suitable for identifying micro-heterogeneities in biopharmaceuticals, particularly monoclonal antibodies, and for fingerprinting chemical and enzymatic alterations during manufacturing, purification, formulation, and storage of monoclonal antibodies.
  • Optimized for creating a gradient for LC/MS analysis of mAb charge variant separations.
  • Ensures high mass spectrum quality.
  • Supplied in one-liter trace metal certified low-density polyethylene bottles.
  • Contains 100 mL aliquots of 10x strength combinations with proven trace ppb values of sodium, potassium, and calcium.
  • Designed to produce a 160 mM ammonium acetate mobile phase with a pH of 8.5.
  • Contains 20% acetonitrile to prevent bacterial growth.
Catalog No. 50-203-8968 Supplier Waters Corp Supplier No. 186009281
This 10X concentrate was developed in conjunction with BioResolve SCX columns and is optimized for creating a gradient for LC/MS analysis of mAb charge variant separations. It is suitable for identifying micro-heterogeneities in biopharmaceuticals, particularly monoclonal antibodies, and for fingerprinting chemical and enzymatic alterations during manufacturing, purification, formulation, and storage of monoclonal antibodies.
  • Optimized for creating a gradient for LC/MS analysis of mAb charge variant separations.
  • Ensures high mass spectrum quality.
  • Supplied in one-liter trace metal certified low-density polyethylene bottles.
  • Contains 100 mL aliquots of 10x strength combinations with proven trace ppb values of sodium, potassium, and calcium.
  • Designed to produce a 160 mM ammonium acetate mobile phase with a pH of 8.5.
  • Contains 20% acetonitrile to prevent bacterial growth.