Learn More
Waters Corp Waters pH Concentrates, A & B Kit, 10X, MS Certified LDPE Containers, 100 mL
Catalog No. NC1860877 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The IonHance CX-MS pH 10X Concentrates A and B are optimized for use in creating a pH 5.0 to 8.5 gradient for LC/MS analysis of mAb charge variant separations. These concentrates are designed to be utilized alongside BioResolve SCX columns. They aim to improve sensitivity, retentivity, and peak shape for challenging separations.
  • Optimized for creating a pH 5.0 to 8.5 gradient for LC/MS analysis.
  • Designed for mAb charge variant separations.
  • Used in conjunction with BioResolve SCX columns.
  • Assists in achieving low metal content and high-quality standards.
  • Improves sensitivity, retentivity, and peak shape for challenging separations.
Catalog No. NC1860877 Supplier Waters Corp Supplier No. 176004498
The IonHance CX-MS pH 10X Concentrates A and B are optimized for use in creating a pH 5.0 to 8.5 gradient for LC/MS analysis of mAb charge variant separations. These concentrates are designed to be utilized alongside BioResolve SCX columns. They aim to improve sensitivity, retentivity, and peak shape for challenging separations.
  • Optimized for creating a pH 5.0 to 8.5 gradient for LC/MS analysis.
  • Designed for mAb charge variant separations.
  • Used in conjunction with BioResolve SCX columns.
  • Assists in achieving low metal content and high-quality standards.
  • Improves sensitivity, retentivity, and peak shape for challenging separations.