Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Mobile-Phase Additive, Difluoroacetic Acid, MS-Grade, 1 mL

Catalog No. NC1724129 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This difluoroacetic acid is an MS-grade mobile-phase additive that can be used for LC and LC-MS techniques, such as reversed-phase separations of proteins, peptides, and small molecules. It has been purified to achieve the low metal (sodium and potassium) content necessary for MS compatibility. Each vial is provided in an approximately 1 mL quantity and is contained within a high quality, chemically resistant PFA vial.

  • MS-grade mobile-phase additive for LC and LC-MS techniques
  • Suitable for reversed-phase separations of proteins, peptides, and small molecules
  • Purified for low metal content (sodium and potassium no greater than 100 ppb) for MS compatibility
  • Volume: 1 mL
  • Formulation: Liquid
  • Storage condition: 4°C
  • Shelf life: 36 months
  • Purity of 98% or higher via GC testing
  • Packaged in a high-quality, chemically resistant PFA vial

Catalog No. NC1724129 Supplier Waters Corp Supplier No. 186009201
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This difluoroacetic acid is an MS-grade mobile-phase additive that can be used for LC and LC-MS techniques, such as reversed-phase separations of proteins, peptides, and small molecules. It has been purified to achieve the low metal (sodium and potassium) content necessary for MS compatibility. Each vial is provided in an approximately 1 mL quantity and is contained within a high quality, chemically resistant PFA vial.

  • MS-grade mobile-phase additive for LC and LC-MS techniques
  • Suitable for reversed-phase separations of proteins, peptides, and small molecules
  • Purified for low metal content (sodium and potassium no greater than 100 ppb) for MS compatibility
  • Volume: 1 mL
  • Formulation: Liquid
  • Storage condition: 4°C
  • Shelf life: 36 months
  • Purity of 98% or higher via GC testing
  • Packaged in a high-quality, chemically resistant PFA vial

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.