Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Mobile Phase Kit, MS-grade, Difluoroacetic Acid, Low Metal Content

Catalog No. NC1735242 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This MS-grade mobile-phase additive for LC and LC-MS aids reversed-phase separations of proteins, peptides, and small molecules. It offers low metal (sodium & potassium) content for MS compatibility. Kit includes two 1.1 mL additive vials and two 1000 mL LDPE mobile phase bottles, rigorously QC tested for <100 ppb sodium & potassium.

  • MS-grade mobile-phase additive for LC & LC-MS
  • Ideal for reversed-phase separations (proteins, peptides, small molecules)
  • Low metal (sodium & potassium) content ensures MS compatibility
  • Each standard vial: approx. 1 mL in chemically resistant PFA
  • Kit: two 1.1 mL additive vials & two 1000 mL LDPE mobile phase bottles
  • Containers QC tested for <100 ppb sodium & potassium
  • Purity >98% via GC testing
  • Shelf life: 36 months, Shipping: Ambient
  • Store at 4 °C ± 4 °C until expiration

Catalog No. NC1735242 Supplier Waters Corp Supplier No. 176010000
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This MS-grade mobile-phase additive for LC and LC-MS aids reversed-phase separations of proteins, peptides, and small molecules. It offers low metal (sodium & potassium) content for MS compatibility. Kit includes two 1.1 mL additive vials and two 1000 mL LDPE mobile phase bottles, rigorously QC tested for <100 ppb sodium & potassium.

  • MS-grade mobile-phase additive for LC & LC-MS
  • Ideal for reversed-phase separations (proteins, peptides, small molecules)
  • Low metal (sodium & potassium) content ensures MS compatibility
  • Each standard vial: approx. 1 mL in chemically resistant PFA
  • Kit: two 1.1 mL additive vials & two 1000 mL LDPE mobile phase bottles
  • Containers QC tested for <100 ppb sodium & potassium
  • Purity >98% via GC testing
  • Shelf life: 36 months, Shipping: Ambient
  • Store at 4 °C ± 4 °C until expiration

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.