Learn More
Waters Corp IonHance Difluoroacetic Acid Super Kit (6 Vials)
Catalog No. NC1641495 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Waters IonHance Difluoroacetic Acid is an MS-grade mobile-phase additive that can be used for LC and LC-MS techniques, such as reversed-phase separations of proteins, peptides, and small molecules. This reagent has been purified to achieve the low metal (sodium and potassium) content that is necessary for MS compatibility. Each vial of the standard is provided in an approximately 1 mL quantity and is contained within a high quality, chemically resistant PFA vial.
Catalog No. NC1641495 Supplier Waters Corp Supplier No. 176004415
Waters IonHance Difluoroacetic Acid is an MS-grade mobile-phase additive that can be used for LC and LC-MS techniques, such as reversed-phase separations of proteins, peptides, and small molecules. This reagent has been purified to achieve the low metal (sodium and potassium) content that is necessary for MS compatibility. Each vial of the standard is provided in an approximately 1 mL quantity and is contained within a high quality, chemically resistant PFA vial.