This high purity MS-grade mobile phase additive is designed for oligonucleotide LC-MS analysis. It is manufactured and purified to achieve the lowest metal content, improving MS signal and spectra quality. Each chemically resistant PFA container includes approximately 10 mL of reagent. It is a weak acid used with an amine reagent to provide high ionization efficiency mobile phases, certified by GC to have a purity ≥99% and contains <100 ppb sodium (Na) and <100 ppb potassium (K) content.
- High purity MS-grade mobile phase additive
- Lowest metal content to improve MS signal and spectra quality
- Approximately 10 mL of reagent in a chemically resistant PFA container
- Used in combination with amine reagent for high ionization efficiency mobile phases
- Certified by GC to have a purity ≥99%
- Contains <100 ppb sodium (Na) and <100 ppb potassium (K) content