Learn More
Hampton Research IONIC LIQUID SCREEN #09
SDP
Catalog No. 50563082
Encompass
HR2-214-09/Ionic Liquid Screen #09, 9 ml - 50% w/v 1-Ethyl-3-methylimidazolium chloride. Please note items from this supplier are non-returnable.
Catalog No. 50-563-082 Supplier Hampton Research Supplier No. HR221409
HR2-214-09/Ionic Liquid Screen #09, 9 ml - 50% w/v 1-Ethyl-3-methylimidazolium chloride. Please note items from this supplier are non-returnable.