Learn More
Hampton Research IONIC LIQUID SCREEN #13
SDP
Catalog No. 50563086
Encompass
HR2-214-13/Ionic Liquid Screen #13, 9 ml - 50% w/v 1,3-Dimethylimidazolium methyl sulfate. Please note items from this supplier are non-returnable.
Catalog No. 50-563-086 Supplier Hampton Research Supplier No. HR221413
HR2-214-13/Ionic Liquid Screen #13, 9 ml - 50% w/v 1,3-Dimethylimidazolium methyl sulfate. Please note items from this supplier are non-returnable.