Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ionic Strength Adjustor, pH Indicating and Adjusting, for use with Ammonia Selective Electrode, Ricca Chemical
SDP

Catalog No. SA0018801A Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SA0018801A 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. SA0018801A Supplier Ricca Chemical Company Supplier No. SA0018801A

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For Use With Ammonia Selective Electrode

Specifications

Chemical Name or Material Ionic Strength Adjustor
Packaging Poly natural
CAS Min % 65.0, 26.25, 5.14, 1.22, 0.01
CAS Max % 68.0, 27.32, 5.35, 1.27, 0.01
CAS 1310-73-2, 67-56-1, 6381-92-6, 125-20-2
Physical Form Liquid
Color Blue
Quantity 1 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.