Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ionic Strength Adjustor, for use with Sodium Ion Selective Electrode, Ricca Chemical
SDP

Catalog No. R41300004A Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R41300004A 4 L
4130-16 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R41300004A Supplier Ricca Chemical Company Supplier No. 41301
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material Ionic Strength Adjustor
Color Colorless
Name Note For use with Sodium Ion Selective Electrode
CAS 7732-18-5, 1336-21-6, 12125-02-9
CAS Min % 68.0, 16.5, 13.59
CAS Max % 71.0, 17.18, 14.15
Grade Laboratory
Packaging Natural Poly Bottle
pH Alkaline
Quantity 4 L
Physical Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.