Learn More
Medchemexpress LLC Ionizable cationic lipid (306-N16B) | 2803699-70-7 | 98.8% | 1471.6 | C75H151N7O4S8 | 10 MG
SDP
Catalog No. 5000286354
Encompass_Preferred
306-N16B is an ionizable cationic lipid used to formulate lipid nanoparticles (LNPs) for targeted mRNA and gene delivery to the lung. It has been reported to preferentially target lung endothelial cells and select immune cell populations, and is supplied for research use with characterized molecular and purity data suitable for formulation and analytical work.
  • Enables pH-dependent endosomal escape for efficient intracellular delivery.
  • Supports selective lung targeting of lipid nanoparticle formulations.
  • Compatible with mRNA and gene delivery workflows.
  • High purity and defined molecular properties for reproducible formulation.
  • Available in small mg-scale quantities for R&D applications.
Catalog No. 50-002-86354 Supplier Medchemexpress LLC Supplier No. HY15022910MG
306-N16B is an ionizable cationic lipid used to formulate lipid nanoparticles (LNPs) for targeted mRNA and gene delivery to the lung. It has been reported to preferentially target lung endothelial cells and select immune cell populations, and is supplied for research use with characterized molecular and purity data suitable for formulation and analytical work.
  • Enables pH-dependent endosomal escape for efficient intracellular delivery.
  • Supports selective lung targeting of lipid nanoparticle formulations.
  • Compatible with mRNA and gene delivery workflows.
  • High purity and defined molecular properties for reproducible formulation.
  • Available in small mg-scale quantities for R&D applications.