Learn More
Mettler Toledo™ Ionizer 1 Point Haug with 6 ft. Cable
Catalog No. 0932042 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Ionizer 1 Pt Haug with 6 ft. Cable
Catalog No.
09-320-42 Ionizer 1 Pt Haug with 6 ft. Cable
1 options
Catalog No. 09-320-42 Supplier Mettler Toledo™ Supplier No. 63052306
Add to Cart
Add to Cart
The ionizer instantaneously neutralizes the sample while keeping the weighing pan free and accessible. Stabilization time of the weighing is not affected because there are no drafts in the weighing chamber.
  • Discharges electrostatically charged samples for precise weighing
  • A high-voltage, low-current charge neutralizes the disruptive electrostatic charge on the sample, eliminating any forces that may distort weighing
  • The systems can be used with any balance and weighing substance
  • The ionizer instantaneously neutralizes the sample while keeping the weighing pan free and accessible
  • Ionizer is safe to the touch
  • Point ionizer is ideal for use with powdery, toxic samples
  • A flexible cable allows the point ionizer to be easily fastened in any position
TRUSTED_SUSTAINABILITY
Specifications
Type Ionizer