Learn More
Amstat Industries Staticmaster 1U400 1" Ionizers
SDP
Catalog No. NC9514344
Encompass
The 1U400 alpha spot ionizers are ideal small profile static eliminators.  When weighing fine powders and fibers, the balance chamber must maintain a stable static free environment to assure accurate results. The problem of  “fly-away” powders and inaccurate weighing is eliminated with the 1U400 microbalance ionizer because they utilize proven alpha ionizing technology. These devices can be easily mounted within the balance chamber, assuring quick and accurate measurements. They are inherently balanced to zero volts - ensuring thorough static removal every time. It is recommended to replace the ionizer every 12 months to ensure optimal performance. Amstat guarantees made to order new devices to ensure our customers get the longest use out of the ionizers. 
Catalog No. NC9514344 Supplier Amstat Industries Supplier No. 1U400
The 1U400 alpha spot ionizers are ideal small profile static eliminators.  When weighing fine powders and fibers, the balance chamber must maintain a stable static free environment to assure accurate results. The problem of  “fly-away” powders and inaccurate weighing is eliminated with the 1U400 microbalance ionizer because they utilize proven alpha ionizing technology. These devices can be easily mounted within the balance chamber, assuring quick and accurate measurements. They are inherently balanced to zero volts - ensuring thorough static removal every time. It is recommended to replace the ionizer every 12 months to ensure optimal performance. Amstat guarantees made to order new devices to ensure our customers get the longest use out of the ionizers. 