Learn More
Amstat Industries Staticmaster 2U500 3" Ionizers
SDP
Catalog No. NC9514343
Encompass
The 2u500 alpha spot ionizers are ideal small profile static eliminators. When weighing fine powders and fibers, the balance chamber must maintain a stable static free environment to assure accurate results. The problem of “fly-away” powders and inaccurate weighing is eliminated with the 2U500 microbalance ionizer because they utilize proven alpha ionizing technology. These devices can be easily mounted within the balance chamber, assuring quick and accurate measurements. They are inherently balanced to zero volts - ensuring thorough static removal every time. Amstat guarantees made to order new devices to ensure our customers get the longest use out of the ionizers.
Catalog No. NC9514343 Supplier Amstat Industries Supplier No. 2U500
This product is regulated by the federal government, and may not be ordered online.
The 2u500 alpha spot ionizers are ideal small profile static eliminators. When weighing fine powders and fibers, the balance chamber must maintain a stable static free environment to assure accurate results. The problem of “fly-away” powders and inaccurate weighing is eliminated with the 2U500 microbalance ionizer because they utilize proven alpha ionizing technology. These devices can be easily mounted within the balance chamber, assuring quick and accurate measurements. They are inherently balanced to zero volts - ensuring thorough static removal every time. Amstat guarantees made to order new devices to ensure our customers get the longest use out of the ionizers.