Learn More
Cayman Chemical IonmycIncalcIum salt 10mg
SDP
Catalog No. 502954085
Encompass_Preferred
A calcium ionophore; induces intracellular calcium mobilization in X. laevis oocytes when used at a concentration of 5 µM
Catalog No. 50-295-4085 Supplier Cayman Chemical Supplier No. 1193210 MG
A calcium ionophore; induces intracellular calcium mobilization in X. laevis oocytes when used at a concentration of 5 µM