Learn More
Abcam Ionomycin Ca2+ Salt, Ca2+ ionophore, 5MG
Catalog No. 502584688
Encompass_Preferred
MW 747.1 Da, Purity >99%. Ca²+ ionophore. Useful when calcium-dose response data are not required. Ion specificity Mn²+>Ca²+>Mg²+>Sr²+>Ba²+.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-4688 Supplier Abcam Supplier No. AB1201165MG
Add to Cart
Add to Cart
MW 747.1 Da, Purity >99%. Ca²+ ionophore. Useful when calcium-dose response data are not required. Ion specificity Mn²+>Ca²+>Mg²+>Sr²+>Ba²+.
The product is subject to the following: Abcam Restricted Use Statement