Learn More
Lkt Laboratories Ionomycin Calcium, ≥98%, 1MG
SDP
Catalog No. NC9966566
Encompass_Preferred
Polyether Ca2+ ionophore. CAS: 56092-82-1. Additional sizes available upon request. Ships from Saint Paul, MN.
Catalog No. NC9966566 Supplier Lkt Laboratories Supplier No. I57531MG
Polyether Ca2+ ionophore. CAS: 56092-82-1. Additional sizes available upon request. Ships from Saint Paul, MN.