Learn More
Medchemexpress LLC HY-13434A 5mg Medchemexpress, Ionomycin (calcium) CAS:56092-82-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13434A5MG
Encompass_Preferred
Medchemexpress, HY-13434A 5mg Ionomycin (calcium) CAS:56092-82-1 Ionomycin calcium (SQ23377 calcium) is a potent, selective calcium ionophore and an antibiotic produced by Streptomyces conglobatus. Ionomycin calcium (SQ23377 calcium) is highly specific for divalent cations (Ca>Mg>Sr=Ba). Ionomycin (SQ23377) promotes apoptosis. Ionomycin calcium (SQ23377 calcium) also induces the activation of protein kinase C (PKC)[2][3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0311