Learn More
AdipoGen Ionomycin Calcium Salt, 56092-82-1, MFCD00083634, 1 mg
SDP
Catalog No. 50596349
Encompass_Preferred
MF: C41H70O9 . Ca, MW: 707.0 . 40.1, ≥98%, Appearance: White to off-white solid. Antibiotic, Potent and highly selective Ca2+ ionophore. Commonly used to modify intracellular calcium levels to study calcium transport across biological membranes and to calibrate fluorescent Ca2+ indicators.
Catalog No. 50-596-349 Supplier AdipoGen Supplier No. AGCN20418M001
MF: C41H70O9 . Ca, MW: 707.0 . 40.1, ≥98%, Appearance: White to off-white solid. Antibiotic, Potent and highly selective Ca2+ ionophore. Commonly used to modify intracellular calcium levels to study calcium transport across biological membranes and to calibrate fluorescent Ca2+ indicators.