Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ionomycin, calcium salt, >98%, MP Biomedicals™

Catalog No. ICN15507005 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15507005 5 mg
ICN15507001 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15507005 Supplier MP Biomedicals, Inc Supplier No. 0215507005
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 56092-82-1 | Formula: C41H70CaO9 | Weight: 747.08 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56092-82-1
Molecular Weight (g/mol) 747.08
MDL Number MFCD00083634
InChI Key RULWQOBWZOQCOY-SQNLDGQHSA-M
PubChem CID 71311740
IUPAC Name calcium;(4R,6S,8S,10E,12R,14R,16E,18R,19R,20S,21S)-19,21-dihydroxy-22-[(2S,5S)-5-[(2R,5S)-5-[(1R)-1-hydroxyethyl]-5-methyloxolan-2-yl]-5-methyloxolan-2-yl]-4,6,8,12,14,18,20-heptamethyl-11-oxido-9-oxodocosa-10,16-dienoate
SMILES [Ca++].C[C@H](CCC([O-])=O)C[C@H](C)C[C@H](C)C(=O)[CH-]C(=O)[C@H](C)C[C@H](C)C\C=C\[C@@H](C)[C@@H](O)[C@@H](C)[C@@H](O)C[C@@H]1CC[C@](C)(O1)[C@H]1CC[C@](C)(O1)[C@@H](C)O

Specifications

Type Ionomycin Calcium Salt
Physical Form Crystalline Powder
Color White
Quantity 5 mg
Assay Percent Range 1
Molecular Formula C41H70CaO9
Formula Weight 747.1
Percent Purity >98%
Grade Not Available
Chemical Name or Material Ionomycin, calcium salt, For HPLC analysis
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.