Learn More
Enzo Life Sciences Ionomycin . calcium salt, (1mg), CAS Number: 56092-82-1
SDP
Catalog No. NC9512743
Encompass_Preferred
Calcium ionophore. For technical information and literature references see Ionomycin (free acid) (Prod. No. ALX-450-006). Formula: C41H70O9. Ca. MW: 707.0. 40.1. CAS Number: 56092-82-1. Purity: ≥97% (HPLC, TLC). Appearance: White to off-white solid. Solubility: 10mg/ml soluble in DMSO or 100% ethanol, 5mg/ml soluble in methanol. Long Term Storage: +4°C
Catalog No. NC9512743 Supplier Enzo Life Sciences Supplier No. ALX450007M001
Calcium ionophore. For technical information and literature references see Ionomycin (free acid) (Prod. No. ALX-450-006). Formula: C41H70O9. Ca. MW: 707.0. 40.1. CAS Number: 56092-82-1. Purity: ≥97% (HPLC, TLC). Appearance: White to off-white solid. Solubility: 10mg/ml soluble in DMSO or 100% ethanol, 5mg/ml soluble in methanol. Long Term Storage: +4°C