Learn More
BIOGEMS INTERNATIONAL INC Ionomycin Calcium Salt 1mg
SDP
Supplier:  BIOGEMS INTERNATIONAL INC 56082121MG
Encompass
Ionomycin is an ionphore discovered in Streptomyces conglobatus and used in the study of intracellular calcium. It is also reported to stimulate the intracellular production of IL-2, IL-4 and perforin. It is supplied as a a calcium salt for ease of storage.
Catalog No. 50-153-0188