Learn More
Biotium Ionomycin
Supplier:  Biotium 59007
Encompass_Preferred
Ionomycin is commonly used to modify intracellular calcium concentrations and to calibrate fluorescent calcium indicators. It is also known for its antiproliferative effects. Properties: Yellowish oily solid soluble in DMSO or EtOH; Store desiccated at < 4°C C41H70O9Ca; MW: 747.1; [56092-82-1].
Catalog No. 50-196-4713