Learn More
Sigma Aldrich Fine Chemicals Biosciences Ionomycin calcium salt, MFCD00083634, 1mL
Catalog No. 501657320
Encompass
Empirical Formula (Hill Notation) C41H70CaO9, Molecular Weight 747.07, Ready Made Solution, from Streptomyces conglobatus, 1 mM in DMSO, 0.2 μm filtered
Catalog No. 50-165-7320 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. I39091ML
Empirical Formula (Hill Notation) C41H70CaO9, Molecular Weight 747.07, Ready Made Solution, from Streptomyces conglobatus, 1 mM in DMSO, 0.2 μm filtered