Learn More
Enzo Life Sciences Ionomycin. calcium salt (5mg). CAS: 56092-82-1
SDP
Catalog No. 502009278
Encompass_Preferred
For technical information and literature references see Ionomycin (free acid) (ALX-450-006). Purity: ≥97% (HPLC, TLC). Solubility: 10mg/ml soluble in DMSO or 100% ethanol, 5mg/ml soluble in methanol. Long Term Storage: +4°C.
Catalog No. 50-200-9278 Supplier Enzo Life Sciences Supplier No. ALX450007M005
For technical information and literature references see Ionomycin (free acid) (ALX-450-006). Purity: ≥97% (HPLC, TLC). Solubility: 10mg/ml soluble in DMSO or 100% ethanol, 5mg/ml soluble in methanol. Long Term Storage: +4°C.