Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ionomycin (Calcium Salt), Fisher BioReagents™

Catalog No. BP25271 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 mg
Packaging:
Poly Micro Tube
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP25271 1 mg Poly Micro Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP25271

missing translation for 'validMainframeMsg'

Ionomycin (Calcium Salt)

  •  Suitable for use in electrophoretic separations, specifically polyacrylamide gel electrophoresis (PAGE)
  • This product is a 40% (w/v) solution of high quality acrylamide
  • Free of DNase, RNase, and protease
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56092-82-1
Molecular Formula C41H70CaO9
Molecular Weight (g/mol) 747.08
MDL Number MFCD00083634
InChI Key RULWQOBWZOQCOY-SQNLDGQHSA-M
PubChem CID 71311740
IUPAC Name calcium;(4R,6S,8S,10E,12R,14R,16E,18R,19R,20S,21S)-19,21-dihydroxy-22-[(2S,5S)-5-[(2R,5S)-5-[(1R)-1-hydroxyethyl]-5-methyloxolan-2-yl]-5-methyloxolan-2-yl]-4,6,8,12,14,18,20-heptamethyl-11-oxido-9-oxodocosa-10,16-dienoate
SMILES [Ca++].C[C@H](CCC([O-])=O)C[C@H](C)C[C@H](C)C(=O)[CH-]C(=O)[C@H](C)C[C@H](C)C\C=C\[C@@H](C)[C@@H](O)[C@@H](C)[C@@H](O)C[C@@H]1CC[C@](C)(O1)[C@H]1CC[C@](C)(O1)[C@@H](C)O

Specifications

Type Ionomycin Calcium Salt
Physical Form Solid
Melting Point 205°C to 206°C
Packaging Poly Micro Tube
Color White
Quantity 1 mg
Assay Percent Range ≥98.0 %
Formula Weight 747.1
Percent Purity 98.0% min. (by NP TLC), 99.0% min. (by RP HPLC)
Chemical Name or Material Ionomycin, Calcium Salt, From Streptomyces conglobatus
Safety handling Check Alert storage code Red
Recommended Storage 2 to 8°C, protect from light
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.