Learn More
AdipoGen Ionomycin (free acid), 56092-81-0, MFCD06798385, 1 mg
SDP
Catalog No. 50596347
Encompass_Preferred
MF: C41H72O9, MW: 709.0, ≥98%, Appearance: Waxy yellow to colorless solid. Antibiotic, Potent and highly selective Ca2+ ionophore. Commonly used to modify intracellular calcium levels to study calcium transport across biological membranes and to calibrate fluorescent Ca2+ indicators.
Catalog No. 50-596-347 Supplier AdipoGen Supplier No. AGCN20416M001
MF: C41H72O9, MW: 709.0, ≥98%, Appearance: Waxy yellow to colorless solid. Antibiotic, Potent and highly selective Ca2+ ionophore. Commonly used to modify intracellular calcium levels to study calcium transport across biological membranes and to calibrate fluorescent Ca2+ indicators.