Learn More
Alkali Scientific Ionomycin Free Acid, 1 Mg
SDP
Catalog No. 5000659613
Encompass_Preferred
Ionomycin is a calcium ionophore used to facilitate the transport of calcium ions across cell membranes. This 1 mg formulation is typically used in cell culture systems to manipulate intracellular calcium levels, which are crucial for various cellular processes such as signaling, gene expression, and apoptosis. By increasing cytosolic calcium levels, ionomycin activates calcium-dependent pathways, providing insights into the role of calcium in cell function. It is widely used in immunology and cell biology research, particularly for studying calcium-dependent processes in T cell activation, muscle contraction, and neuronal signaling.
Catalog No. 50-006-59613 Supplier Alkali Scientific Supplier No. ION003CP
Add to Cart
Add to Cart
Ionomycin is a calcium ionophore used to facilitate the transport of calcium ions across cell membranes. This 1 mg formulation is typically used in cell culture systems to manipulate intracellular calcium levels, which are crucial for various cellular processes such as signaling, gene expression, and apoptosis. By increasing cytosolic calcium levels, ionomycin activates calcium-dependent pathways, providing insights into the role of calcium in cell function. It is widely used in immunology and cell biology research, particularly for studying calcium-dependent processes in T cell activation, muscle contraction, and neuronal signaling.