Learn More
Enzo Life Sciences Ionomycin (free acid) (1mg). CAS: 56092-81-0
SDP
Catalog No. 502009276
Encompass_Preferred
Potent and highly selective calcium ionophore. Ca2+/ionomycin complex forms between pH 7-9.5 which has intense UV absorption. Induces apoptotic neuronal degeneration in embryonic cortical neurons and cell cycle arrest at G1 phase. Useful tool for increasing intracellular Ca2+ concentration. More effective than A23187. Non fluorescent. Has been used at 4nM and 2-4µM  (whole cells). Purity: ≥90% (HPLC, TLC). Appearance: Waxy yellow to colorless material. Solubility: Soluble in DMSO or methanol. Long Term Storage: +4°C.
Catalog No. 50-200-9276 Supplier Enzo Life Sciences Supplier No. ALX450006M001
Potent and highly selective calcium ionophore. Ca2+/ionomycin complex forms between pH 7-9.5 which has intense UV absorption. Induces apoptotic neuronal degeneration in embryonic cortical neurons and cell cycle arrest at G1 phase. Useful tool for increasing intracellular Ca2+ concentration. More effective than A23187. Non fluorescent. Has been used at 4nM and 2-4µM  (whole cells). Purity: ≥90% (HPLC, TLC). Appearance: Waxy yellow to colorless material. Solubility: Soluble in DMSO or methanol. Long Term Storage: +4°C.