Learn More
Enzo Life Sciences Ionomycin (free acid) (5mg). CAS: 56092-81-0
SDP
Supplier:  Enzo Life Sciences ALX450006M005
Encompass_Preferred
Potent and highly selective calcium ionophore. Ca2+/ionomycin complex forms between pH 7-9.5 which has intense UV absorption. Induces apoptotic neuronal degeneration in embryonic cortical neurons and cell cycle arrest at G1 phase. Useful tool for increasing intracellular Ca2+ concentration. More effective than A23187. Non fluorescent. Has been used at 4nM and 2-4µM  (whole cells). Purity: ≥90% (HPLC, TLC). Solubility: Soluble in DMSO or methanol. Long Term Storage: +4°C.
Catalog No. 50-200-9277