Learn More
Apexbio Technology LLC Ionomycin free acid(Synonyms: Ionomycin (free acid), Antibiotic SQ 23377, SQ 23377, Calcium ionophore ionomycin), 5mg, CAS: 56092-81-0.
SDP
Catalog No. 502240037
Encompass_Preferred
Ionomycin free acid (CAS 56092-81-0) is a selective calcium ionophore functioning by mediating Ca transport across biological membranes It forms complexes with calcium ions facilitating their passage through lipid bilayers and increasing intracellular Ca concentrations In vitro studies demonstrate its capacity to transfer calcium ions from aqueous to organic phases highlighting its ion carrier function In vivo experiments indicate ionomycin effectively activates mammalian oocytes promoting subsequent embryonic development Clinical research has employed ionomycin to assess improvements in fertilization outcomes for patients with decreased ovarian reserves
This item is not returnable. View return policy
Catalog No. 50-224-0037 Supplier Apexbio Technology LLC Supplier No. B69475
Ionomycin free acid (CAS 56092-81-0) is a selective calcium ionophore functioning by mediating Ca transport across biological membranes It forms complexes with calcium ions facilitating their passage through lipid bilayers and increasing intracellular Ca concentrations In vitro studies demonstrate its capacity to transfer calcium ions from aqueous to organic phases highlighting its ion carrier function In vivo experiments indicate ionomycin effectively activates mammalian oocytes promoting subsequent embryonic development Clinical research has employed ionomycin to assess improvements in fertilization outcomes for patients with decreased ovarian reserves