Learn More
Sigma Aldrich Fine Chemicals Biosciences Ionomycin, Free Acid, Streptomyces conglobatus, 1mg
Catalog No. 502305803
Encompass
Ionomycin, Free Acid, Streptomyces conglobatus, an ionophore with high specificity for divalent cations, 56092-81-0, 1mg
Catalog No. 50-230-5803 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 4079501MG
Ionomycin, Free Acid, Streptomyces conglobatus, an ionophore with high specificity for divalent cations, 56092-81-0, 1mg