Learn More
Enzo Life Sciences SCREEN-WELL Ionotropic Glutamatergic ligand library (100µl/well)
SDP
Catalog No. 502010200
Encompass_Preferred
The Ionotropic Glutamatergic Ligands Plate contains 60 ligands including endogenous neurotransmitters, agonists, antagonists, and marketed drugs. The library is ideal for screening or identifying recombinant orphan G protein coupled receptors, target validation, secondary screening, assay development, and other pharmacological applications. The Ionotropic Glutamatergic Ligands plate is 1 plate of the larger Neurotransmitter Library (BML-2810) which contains over 680 ligands. Long Term Storage: -80°C. Applications: HTS .
Catalog No. 50-201-0200 Supplier Enzo Life Sciences Supplier No. BML28170100
The Ionotropic Glutamatergic Ligands Plate contains 60 ligands including endogenous neurotransmitters, agonists, antagonists, and marketed drugs. The library is ideal for screening or identifying recombinant orphan G protein coupled receptors, target validation, secondary screening, assay development, and other pharmacological applications. The Ionotropic Glutamatergic Ligands plate is 1 plate of the larger Neurotransmitter Library (BML-2810) which contains over 680 ligands. Long Term Storage: -80°C. Applications: HTS .