Learn More
Enzo Life Sciences SCREEN-WELL Ionotropic Glutamatergic ligand library (500 µl/well)
SDP
Supplier:  Enzo Life Sciences BML28170500
Encompass_Preferred
The Ionotropic Glutamatergic Ligands Plate contains 60 ligands including endogenous neurotransmitters, agonists, antagonists, and marketed drugs. The library is ideal for screening or identifying recombinant orphan G protein coupled receptors, target validation, secondary screening, assay development, and other pharmacological applications. The Ionotropic Glutamatergic Ligands plate is 1 plate of the larger Neurotransmitter Library (BML-2810) which contains over 680 ligands. Long Term Storage: -80°C. Applications: HTS .
Catalog No. 50-201-0201