Learn More
Thermo Scientific IONPAC COLUMN CS17 4MM
Catalog No. NC9048878 Shop All Thermo Scientific Products
Encompass
IonPac CS17 Cation-Exchange Column; Thermo Scientific; Dionex; For gradient separation of amine; w/Biogenic Amine, Alkylamine, Diamine; Analytical column: 2 x 250mm, 4 x 250mm; Guard column: 2 x 50mm, 4 x 50mm, Max. operating pressure: 40
Catalog No. NC9048878 Supplier Thermo Scientific Supplier No. 060557
IonPac CS17 Cation-Exchange Column; Thermo Scientific; Dionex; For gradient separation of amine; w/Biogenic Amine, Alkylamine, Diamine; Analytical column: 2 x 250mm, 4 x 250mm; Guard column: 2 x 50mm, 4 x 50mm, Max. operating pressure: 40