Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Dionex™ IonPac™ NG1-10μm Guard Column (2 x 50 mm)
GREENER_CHOICE

Catalog No. 03050853 Shop All Thermo Scientific Products
Change view
Click to view available options
Column Type:
Guard
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Column Type
03-050-853 Guard
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03-050-853 Supplier Thermo Scientific™ Supplier No. 088763
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Column Format: Guard Column; Column Phase: Reversed Phase, Ion-Pairing, and Ion Suppression

Achieve high-efficiency separations of hydrophobic, ionizable compounds with Thermo Scientific™ Dionex™ IonPac™ NS1 10μm Analytical & Guard Columns. The columns of choice for ion-pair or ion-suppression chromatography, these polymer-based, reversed-phase columns provide excellent resolution and peak shape. Packing is a neutral, macroporous, high surface area, ethylvinylbenzene polymer cross linked with 55% divinylbenzene, making IonPac NS1 columns resistant to solvents, acids and bases while permitting the use of eluent from pH 0 to 14.

Polymer-Based Columns for Efficient Reversed-Phase Separations
  • Ideal for separation of large molecules that carry localized charges, such as surfactants
  • Can be used for traditional polymeric reversed-phase applications
  • Use with ion-pair chromatography for difficult separations
  • Available in 5 and 10μm particle sizes
Key Applications

Use IonPac NS1 columns for reliable separations of various compounds: High molecular weight aliphatic, carboxylic acids; Alkyl and aromatic sulfates and sulfonates; Quaternary ammonium ions; Water-soluble vitamins; Sulfur oxides; Metal cyanide complexes; Phenols

TRUSTED_SUSTAINABILITY

Specifications

Column Type Guard
Flow Rate 1 mL/min
For Use With (Application) Reversed Phase, Ion-Pairing, Ion-Suppression
Max. Pressure 300 psi (20 bar)
Particle Size 10 μm
Product Line ProPac
Stationary Phase Divinylbenzene
Type Mixed Mode
Format Microbore
Packing Material Macroporous Ethylvinylbenzene Polymer
pH 0 to 14
Length (Metric) 50 mm
Diameter (Metric) 2 mm
Green Features FSC
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.