Learn More
Medchemexpress LLC Iopamidol | 60166-93-0 | 25 MG

Supplier: Medchemexpress LLC 25MGHYB0684
Iopamidol is a nonionic, X-Ray iodinated contrast agent used for a wide range of diagnostic applications. It contains amide and hydroxyl functionalities that can be exploited for the generation of chemical exchange saturation transfer (CEST) contrast. This product serves as an analytical and reference standard, commonly used in qualitative, quantitative, and methodological research experiments.
- Analytical and reference standard
- Used for X-Ray iodinated contrast
- Supports chemical exchange saturation transfer (CEST) contrast generation
- Suitable for qualitative, quantitative, and methodological research experiments
- Applicable in techniques such as HPLC, GC, and MS
- Can be used for pH mapping of kidneys
- Molecular formula: C17H22I3N3O8
- Molecular weight: 777.09
- Appears as a solid
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.