Learn More
Medchemexpress LLC Iopamidol | 60166-93-0 | 99.9% | 100 MG
SDP
Catalog No. 5000408518
Encompass_Preferred
Iopamidol is a nonionic, X-Ray iodinated contrast agent (CA) primarily used for various diagnostic applications. It features amide and hydroxyl functionalities, which can be leveraged to generate chemical exchange saturation transfer (CEST) contrast. This product is intended for research use only.
  • Nonionic, X-Ray iodinated contrast agent
  • Utilized for various diagnostic applications
  • Contains amide and hydroxyl functionalities
  • Can generate chemical exchange saturation transfer (CEST) contrast
Catalog No. 50-004-08518 Supplier Medchemexpress LLC Supplier No. HYB0684100MG
Iopamidol is a nonionic, X-Ray iodinated contrast agent (CA) primarily used for various diagnostic applications. It features amide and hydroxyl functionalities, which can be leveraged to generate chemical exchange saturation transfer (CEST) contrast. This product is intended for research use only.
  • Nonionic, X-Ray iodinated contrast agent
  • Utilized for various diagnostic applications
  • Contains amide and hydroxyl functionalities
  • Can generate chemical exchange saturation transfer (CEST) contrast