Learn More
Medchemexpress LLC HY-B1664 100mg Medchemexpress, Iopanoic acid CAS:96-83-3 Purity:>98%
SDP
Catalog No. 501932986
Encompass_Preferred
Medchemexpress, HY-B1664 100mg Iopanoic acid CAS:96-83-3 Iopanoic acid is an inhibitor of 5'-Deiodinase and also an iodinated contrast medium. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2986 Supplier Medchemexpress LLC Supplier No. HYB1664100MG
Medchemexpress, HY-B1664 100mg Iopanoic acid CAS:96-83-3 Iopanoic acid is an inhibitor of 5'-Deiodinase and also an iodinated contrast medium. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.