Learn More
Beckman Coulter Beta Mark TCR Vbeta Repertoire Kit, 25 Tests, RUO
Catalog No. NC1016014
Encompass_Preferred
The IOTest Beta Mark Kit is a multi parametric analysis tool designed for quantitative determination of the TCR Vbeta repertoire of human T lymphocytes by flow cytometry.
Catalog No. NC1016014 Supplier Beckman Coulter Supplier No. IM3497
The IOTest Beta Mark Kit is a multi parametric analysis tool designed for quantitative determination of the TCR Vbeta repertoire of human T lymphocytes by flow cytometry.