Learn More
Medchemexpress LLC IOWH-032 | 1191252-49-9 | 99.6% | 545.18 | 500 MG
SDP
Catalog No. 5000387723
Encompass_Preferred
IOWH-032 is a synthetic anti-secretory molecule and a potent CFTR inhibitor with an IC50 value of 8 μM. It also functions as an anti-diarrheal agent, making it suitable for cholera, secretory diarrhea, and general diarrhea research.
  • Potent CFTR inhibitor with an IC50 value of 8 μM
  • Functions as an anti-diarrheal agent
  • Increases ACE-2 expression in SARS-CoV-2 infected CFBE41o-WT cells
  • Suitable for in vitro and in vivo studies
  • Available with detailed datasheet and safety information
Catalog No. 50-003-87723 Supplier Medchemexpress LLC Supplier No. HY18337500MG
Add to Cart
Add to Cart
IOWH-032 is a synthetic anti-secretory molecule and a potent CFTR inhibitor with an IC50 value of 8 μM. It also functions as an anti-diarrheal agent, making it suitable for cholera, secretory diarrhea, and general diarrhea research.
  • Potent CFTR inhibitor with an IC50 value of 8 μM
  • Functions as an anti-diarrheal agent
  • Increases ACE-2 expression in SARS-CoV-2 infected CFBE41o-WT cells
  • Suitable for in vitro and in vivo studies
  • Available with detailed datasheet and safety information