Learn More
Medchemexpress LLC HY-12304 5mg Medchemexpress, IOX1 CAS:5852-78-8 Purity:>98%
SDP
Catalog No. 502081004
Encompass_Preferred
Medchemexpress, HY-12304 5mg IOX1 CAS:5852-78-8 IOX1, 5-Carboxy-8-hydroxyquinoline, is a potent broad‐spectrum inhibitor of 2OG oxygenases, including the JmjC demethylases. IOX1 inhibits KDM4C, KDM4E, KDM2A, KDM3A and KDM6B with IC50 values of 0.6 μM, 2.3 μM, 1.8 μM, 0.1 μM and 1.4 μM, respectively. IOX1 also inhibits ALKBH5. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1004 Supplier Medchemexpress LLC Supplier No. HY123045MG
Medchemexpress, HY-12304 5mg IOX1 CAS:5852-78-8 IOX1, 5-Carboxy-8-hydroxyquinoline, is a potent broad‐spectrum inhibitor of 2OG oxygenases, including the JmjC demethylases. IOX1 inhibits KDM4C, KDM4E, KDM2A, KDM3A and KDM6B with IC50 values of 0.6 μM, 2.3 μM, 1.8 μM, 0.1 μM and 1.4 μM, respectively. IOX1 also inhibits ALKBH5. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.