Learn More
Selleck Chemical LLC IOX2 10mM/1mL 931398-72-0 JICL38
SDP
Supplier:  Selleck Chemical LLC S291910MM/1ML
Encompass_Preferred
IOX2 (JICL38) is a potent inhibitor of HIF-1? prolyl hydroxylase-2 (PHD2) with IC50 of 21 nM in a cell-free assay, >100-fold selectivity over JMJD2A, JMJD2C, JMJD2E, JMJD3, or the 2OG oxygenase FIH. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-3210